Human Angiotensin 2
Supplier: Thermo Scientific Chemicals
J60866-LB0
J60866-MB
CAAAJ60866-LB0EA
150.68
CAD
CAAAJ60866-LB0
CAAAJ60866-MB
Human Angiotensin 2
Proteins and Peptides
Powder
CAS No.: 4474-91-3
Molecular Formula: C50H71N13O12
Formula Weight: 1046.19
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
MDL No.: MFCD00133092
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...