Phosphate, 0.5M buffer soln., pH 6.5, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J60845-AK
J60845-AP
CAAAJ60845-AKEA
194.31
CAD
CAAAJ60845-AK
CAAAJ60845-AP
Phosphate, 0.5M buffer soln., pH 6.5, Thermo Scientific
Buffers
Biological Buffers
Liquid
CAS No.: 7558-80-7
Physical Form: Liquid
MDL No.: MFCD00003527
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...