Nitrendipine

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Ethyl 5-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate

J60841-03 J60841-MC J60841-ME
CAAAJ60841-03EA 1739.64 CAD
CAAAJ60841-03 CAAAJ60841-MC CAAAJ60841-ME
Nitrendipine
Nitrendipine

L-type calcium channel blocker


Powder


Applications: L-type calcium channel blocker

Formula: C₁₈H₂₀N₂O₆
MW: 360.366 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00082255
CAS Number: 39562-70-4
EINECS: 254-513-1
Merck Index: 14,6576

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR