SSC buffer (20X), Thermo Scientific
Supplier: Thermo Scientific Chemicals
J60839-K2
J60839-K3
J60839-K7
CAAAJ60839-K2EA
83.87
CAD
CAAAJ60839-K2
CAAAJ60839-K3
CAAAJ60839-K7
SSC buffer (20X), Thermo Scientific
Buffers
Electrophoresis Buffers
Hybridisation Buffers
Liquid
Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD00132371
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...