Folic acid, crystalline

Supplier: Thermo Scientific Chemicals

J60833-14 J60833-22
CAAAJ60833-14EA 183.71 CAD
CAAAJ60833-14 CAAAJ60833-22
Folic acid, crystalline
Folic acid

Vitamin B9. A B-complex vitamin necessary for the body to produce red blood cells


Crystalline


Vitamin B9. A B-complex vitamin necessary for the body to produce red blood cells

Formula: C₁₉H₁₉N₇O₆
Storage Temperature: Ambient
MDL Number: MFCD00079305
CAS Number: 59-30-3
EINECS: 200-419-0
Merck Index: 14,4221

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR