myo-Inositol 99%

Supplier: Thermo Scientific Chemicals

J60828-22 J60828-36
CAAAJ60828-22EA 169.91 CAD
CAAAJ60828-22 CAAAJ60828-36
myo-Inositol 99%
myo-Inositol

A lipotropic agent


Crystalline powder


A lipotropic agent

Formula: C₆H₁₂O₆
Boiling Pt: 291 °C (1013 hPa)
Melting Pt: 222…227 °C
Density: 1.752 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00077932
CAS Number: 87-89-8
EINECS: 201-781-2
Merck Index: 14,4978

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR