(-)-Epicatechin gallate

Supplier: Thermo Scientific Chemicals

J60814-FPL J60814-MA
CAAAJ60814-FPLEA 757.09 CAD
CAAAJ60814-FPL CAAAJ60814-MA
(-)-Epicatechin gallate
(-)-Epicatechin gallate

A natural product from green tea that induces apoptosis


Powder


A natural product from green tea that induces apoptosis

Formula: C₂₂H₁₈O₁₀
Storage Temperature: Refrigerator
MDL Number: MFCD00075936
CAS Number: 1257-08-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR