Tricine-Buffered Saline (5X), pH 7.0, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J60811-AK
J60811-AP
CAAAJ60811-AKEA
200.11
CAD
CAAAJ60811-AK
CAAAJ60811-AP
Tricine-Buffered Saline (5X), pH 7.0, Thermo Scientific
Buffers
Biological Buffers
Liquid
CAS No.: 5704-04-1
Physical Form: Liquid
Appearance: Colorless
Odor: Odorless
MDL No.: MFCD00004277
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...