Borate, 0.5 M Buffer Solutions, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J60803-AK J60803-AP J62125-AK J62125-AP J62154-AP J63742-AK J63742-AP
CAAAJ60803-AKEA 150.01 CAD
CAAAJ60803-AK CAAAJ60803-AP CAAAJ62125-AK CAAAJ62125-AP CAAAJ62154-AP CAAAJ63742-AK CAAAJ63742-AP
Borate, 0.5 M Buffer Solutions, Thermo Scientific
Buffers Biological Buffers

Liquid


CAS No.: 1330-43-4
MDL No.: MFCD00163147


Caution: Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR