Phosphate-buffered saline (PBS, 10X), pH 7.4, for Western blot, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J60801-K2 J60801-K3 J60801-K7
CAAAJ60801-K2EA 105.23 CAD
CAAAJ60801-K2 CAAAJ60801-K3 CAAAJ60801-K7
Phosphate-buffered saline (PBS, 10X), pH 7.4, for Western blot, Thermo Scientific
Buffers Electrophoresis Buffers Washing Buffers

Liquid


Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD00131855
Applications: For western blot washing.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR