Miconazole nitrate ≥98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (±)-Miconazole nitrate salt, 1-[2-(2,4-Dichlorobenzyloxy)-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole Nitrate

J60796-06 J60796-14
CAAAJ60796-06EA 300.14 CAD
CAAAJ60796-06 CAAAJ60796-14
Miconazole nitrate ≥98%
Miconazole nitrate

Inhibits cytochrome P450-dependent 14a-demethylase


Powder


Applications: Inhibits cytochrome P450-dependent 14a-demethylase

Formula: C₁₈H₁₄Cl₄N₂O·HNO₃
MW: 479.14 g/mol
Melting Pt: 170…185 °C
Storage Temperature: Ambient
MDL Number: MFCD00058161
CAS Number: 22832-87-7
EINECS: 245-256-6
Merck Index: 14,6178

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR