CGS 21680 hydrochloride 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 3-(4-(2-((6-Amino-9-((2R,3R,4S,5S)-5-(ethylcarbamoyl)-3,4-dihydroxytetrahydrofuran-2-yl)-9H-purin-2-yl)amino)ethyl)phenyl)propanoic acid hydrochloride

J60793-LB0 J60793-MA
CAAAJ60793-LB0EA 596.81 CAD
CAAAJ60793-LB0 CAAAJ60793-MA
CGS 21680 hydrochloride 99%
CGS 21680 hydrochloride

An A2A Adenosine receptor agonist. CGS 21680 hydrochloride is currently used in research in studying neuronal transmission, also in respiration.


  • Solid


Solid


CGS 21680 is commonly used to study the actions of the adenosine A2A receptor in cells and tissues. CGS 21680 HCl is an adenosine A2 receptor agonist with IC50 of 22 nM, exhibits 140-fold over A1 receptor. CGS 21680C effectively increases coronary flow with an ED25 value of 1.8 nM. In hippocampal slices, CGS 21680 apis weak agonist on pre- and postsynaptic measures of electrophysiological activity.


Caution: Store away from strong oxidizing agents.

Formula: C₂₃H₂₉N₇O₆·HCl
Storage Temperature: Ambient
MDL Number: MFCD00078573
CAS Number: 124431-80-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR