Guanidinium hydrochloride 99+%

Supplier: Thermo Scientific Chemicals
Warning

J60786-22 J60786-36 J60786-A1
CAAAJ60786-22EA 140.76 CAD
CAAAJ60786-22 CAAAJ60786-36 CAAAJ60786-A1
Guanidinium hydrochloride 99+%
Guanidine hydrochloride

Powder


Purity: 99+% CAS No.: 50-01-1 Molecular Formula: CH5N3.HCl Formula Weight: 95.53 Physical Form: Powder Density: 1.344 Appearance: White Melting Point: 185-188° Solubility: Freely soluble in water Beilstein Reference: 3591990 Merck Reference: 14,4562 MDL No.: MFCD00013026

Formula: NH₂C(=NH)NH₂·HCl
Melting Pt: 185…188 °C
Density: 1.344
Storage Temperature: Ambient
MDL Number: MFCD00013026
CAS Number: 50-01-1
EINECS: 200-002-3
Merck Index: 14,4562
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da5a871cab977e6d9dc666b457732eb1e869a714&vid=b0d4fcb044b6106fce8e40448dbf2ce6e7077ad7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR