D(-)-Glutamine 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: H-D-Gln-OH, (R)-2,5-Diamino-5-oxopentanoic acid, (R)-(-)-Glutamine

J60784-03 J60784-06 J60784-14
CAAAJ60784-03EA 75.33 CAD
CAAAJ60784-03 CAAAJ60784-06 CAAAJ60784-14
D(-)-Glutamine 99+%
D(-)-Glutamine

Kinase inhibitor; has antivasospastic properties


Powder


Kinase inhibitor; has antivasospastic properties

Formula: C₅H₁₀N₂O₃
MW: 146.15
Storage Temperature: Ambient
MDL Number: MFCD00065607
CAS Number: 5959-95-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR