EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid) 0.5 M in aqueous solution pH 8.0
Supplier: Thermo Scientific Chemicals
Synonyms:
EGTA, Ethylenebis(oxyethylenenitrilo)tetra(acetic acid), Kestranal® FE PA, Idranal® VI, Ethylene glycol-O,O'-bis(2-aminoethyl)-N,N,N',N'-tetraacetic acid, Ethylenebis(oxyethylenenitrilo)tetraacetic acid, Ethylene Glycol Bis(2-aminoethyl Ether)-N,N,N,N-tetraacetic acid, 3,12-Bis(carboxymethyl)-6,9-dioxa-3,12-diazatetradecanedioic acid, Titriplex® VI
Liquid
CAS No.: 67-42-5
Molecular Formula: C14H24N2O10
Formula Weight: 380.35
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD00004291
Formula:
C₁₄H₂₄N₂O₁₀ MW: 380.35 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00004291 CAS Number: 67-42-5 Merck Index: 13,03560 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...