NP-40 lysis buffer, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J60766-AK J60766-AP
CAAAJ60766-AKEA 125.07 CAD
CAAAJ60766-AK CAAAJ60766-AP
NP-40 lysis buffer, Thermo Scientific
Buffers Extraction Buffers

Liquid


Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR