TRIS-buffered saline (TBS, 10X) pH 7.4, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J60764-K2 J60764-K3 J60764-K7
CAAAJ60764-K2EA 100.03 CAD
CAAAJ60764-K2 CAAAJ60764-K3 CAAAJ60764-K7
TRIS-buffered saline (TBS, 10X) pH 7.4, Thermo Scientific
Buffers Electrophoresis Buffers Washing Buffers

Liquid


Physical Form: Liquid
Odor: Odorless
MDL No.: MFCD00132476

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR