MES, 1.0 M Buffer Solutions, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J60763-AK
J60763-AP
J61587-AK
J61587-AP
J61656-AK
J61656-AP
J61960-AK
J61960-AP
J62231-AK
J62231-AP
J62752-AK
J62752-AP
J63341-AK
J63341-AP
CAAAJ60763-AKEA
275.17
CAD
CAAAJ60763-AK
CAAAJ60763-AP
CAAAJ61556-AK
CAAAJ61556-AP
CAAAJ61587-AK
CAAAJ61587-AP
CAAAJ61656-AK
CAAAJ61656-AP
CAAAJ61960-AK
CAAAJ61960-AP
CAAAJ62231-AK
CAAAJ62231-AP
CAAAJ62720-AK
CAAAJ62720-AP
CAAAJ62752-AK
CAAAJ62752-AP
CAAAJ63341-AK
CAAAJ63341-AP
MES, 1.0 M Buffer Solutions, Thermo Scientific
Buffers
Biological Buffers
4-Morpholineethanesulfonic acid, monohydrate (MES) buffers solutions.
CAS No.: 145224-94-8
MDL No.: MFCD00283848
Caution: Incompatible with strong oxidizing agents.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...