HEPES-Buffered Saline, 5X Solutions, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J60753-AK
J60753-AP
J61239-AK
J61239-AP
CAAAJ60753-AKEA
150.01
CAD
CAAAJ60753-AK
CAAAJ60753-AP
CAAAJ61239-AK
CAAAJ61239-AP
HEPES-Buffered Saline, 5X Solutions, Thermo Scientific
Buffers
Biological Buffers
Liquid
CAS No.: 7365-45-9
Odor: Odorless
MDL No.: MFCD00006158
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...