Fasudil hydrochloride ≥99%

Supplier: Thermo Scientific Chemicals
Warning

J60751-03 J60751-MD J60751-ME
CAAAJ60751-03EA 586.69 CAD
CAAAJ60751-03 CAAAJ60751-MD CAAAJ60751-ME
Fasudil hydrochloride ≥99%
Fasudil hydrochloride

Powder


Purity: 99+% CAS No.: 105628-07-7 Molecular Formula: C14H17N3O2S.HCl Formula Weight: 327.83 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: White to off-white MDL No.: MFCD00943198

Formula: C₁₄H₁₇N₃O₂S
Storage Temperature: Freezer
MDL Number: MFCD00943198
CAS Number: 105628-07-7
Merck Index: 14,3942

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR