Enalapril
Supplier: Thermo Scientific Chemicals
J60750-03
J60750-06
CAAAJ60750-03EA
276.09
CAD
CAAAJ60750-03
CAAAJ60750-06
Enalapril
Enalapril
Long-acting ACE inhibitor and antihypertensive
Powder
Applications: Long-acting ACE inhibitor and antihypertensive
Formula:
C₂₀H₂₈N₂O₅ Melting Pt: 142…147 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00865774 CAS Number: 75847-73-3 Merck Index: 14,3567 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...