Thymidine-5'-monophosphate disodium salt
Supplier: Thermo Scientific Chemicals
Synonyms:
5′-Thymidylic acid disodium salt, TMP disodium salt, Sodium ((2R,3S,5R)-3-hydroxy-5-(5-methyl-2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)tetrahydrofuran-2-yl)methyl phosphate, dTMP disodium salt, Thymidine 5′-monophosphate disodium salt hydrate
Powder
CAS No.: 33430-62-5
Molecular Formula: C10H13N2Na2O8P
Formula Weight: 366.18
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Solubility: Soluble in water
Beilstein Reference: 4086927
MDL No.: MFCD09039259
Formula:
C₁₀H₁₃N₂Na₂O₈P Storage Temperature: Ambient |
MDL Number:
MFCD09039259 CAS Number: 33430-62-5 EINECS: 251-518-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...