Ponceau S for electrophoresis

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Hydroxy-4-(2-sulfo-4-[4-sulfophenylazo]phenylazo)-2,7-naphthalenedisulfonic acid sodium salt, 3-Hydroxy-4-(2-sulpho-4-[4-sulphophenylazo]phenylazo)-2,7-naphthalenedisulphonic acid sodium salt, CI 27195, 3-Hydroxy-4-((2-sulfo-4-((4-sulfophenyl)diazenyl)phenyl)diazenyl)naphthalene-2,7-disulfonic acid, tetrasodium salt, Ponceau, 3-Hydroxy-4-((2-sulpho-4-((4-sulphophenyl)diazenyl)phenyl)diazenyl)naphthalene-2,7-disulphonic acid, tetrasodium salt

J60744-09 J60744-18 J60744-30
CAAAJ60744-09EA 90.12 CAD
CAAAJ60744-09 CAAAJ60744-18 CAAAJ60744-30
Ponceau S for electrophoresis
Ponceau S

Protein stain for electrophoresis.


Powder


Protein stain for electrophoresis

Formula: C₂₂H₁₂N₄Na₄O₁₃S₄
MW: 760.56 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003892
CAS Number: 6226-79-5
EINECS: 228-319-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR