Irinotecan hydrochloride
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(S)-4,11-Diethyl-4-hydroxy-3,14-dioxo-3,4,12,14-tetrahydro-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinolin-9-yl [1,4'-bipiperidine]-1'-carboxylate hydrochloride
J60743-MD
J60743-MF
CAAAJ60743-MDEA
1437.34
CAD
CAAAJ60743-MD
CAAAJ60743-MF
Irinotecan hydrochloride
Irinotecan hydrochloride
Topoisomerase I inhibitor
Yellow powder
Applications: Topoisomerase I inhibitor
Formula:
C₃₃H₃₈N₄O₆·HCl Storage Temperature: Freezer |
MDL Number:
MFCD01862255 CAS Number: 100286-90-6 Merck Index: 14,5091 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...