2'-Deoxyguanosine monohydrate

Supplier: Thermo Scientific Chemicals

J60741-03 J60741-06 J60741-09
CAAAJ60741-03EA 200.11 CAD
CAAAJ60741-03 CAAAJ60741-06 CAAAJ60741-09
2'-Deoxyguanosine monohydrate
2'-Deoxyguanosine monohydrate

Crystalline powder


CAS No.: 961-07-9 Molecular Formula: C10H13N5O4 Formula Weight: 267.25 Physical Form: Crystalline powder Appearance: White to off-white

Formula: C₁₀H₁₃N₅O₄·H₂O
Storage Temperature: Ambient
MDL Number: MFCD00150760
CAS Number: 312693-72-4
EINECS: 213-505-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR