Glycylglycine, 0.2 M buffer soln., pH 8.5, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J60717-AE
J60717-AK
CAAAJ60717-AEEA
88.26
CAD
CAAAJ60717-AE
CAAAJ60717-AK
Glycylglycine, 0.2 M buffer soln., pH 8.5, Thermo Scientific
Buffers
Biological Buffers
Liquid
CAS No.: 556-50-3
Physical Form: Liquid
Appearance: Colorless
Odor: Odorless
MDL No.: MFCD00008130
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...