α-Cyclodextrin 97+%

Supplier: Thermo Scientific Chemicals

J60687-06 J60687-14 J60687-22
CAAAJ60687-06EA 100.9 CAD
CAAAJ60687-06 CAAAJ60687-14 CAAAJ60687-22
α-Cyclodextrin 97+%
α-Cyclodextrine

Useful for selective precipitation of enantiomeric, positional or structural isomers


Powder


Useful for selective precipitation of enantiomeric, positional or structural isomers

Formula: C₃₆H₆₀O₃₀
MW: 972.84 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00078207
CAS Number: 10016-20-3
EINECS: 233-007-4
Merck Index: 14,2718

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR