GBR 12909 dihydrochloride

Supplier: Thermo Scientific Chemicals

J60684-MA J60684-MF
CAAAJ60684-MAEA 358.64 CAD
CAAAJ60684-MA CAAAJ60684-MF
GBR 12909 dihydrochloride
GBR 12909 dihydrochloride

Dopamine uptake inhibitor


Powder


Soluble in DMSO to 50mM. Soluble to 5mM in water with warming.

Formula: C₂₈H₃₂F₂N₂O·2HCl
Storage Temperature: Ambient
MDL Number: MFCD00055193
CAS Number: 67469-78-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR