Hydroxytacrine maleate salt
Supplier: Thermo Scientific Chemicals
J60680-03
J60680-MD
CAAAJ60680-03EA
600.12
CAD
CAAAJ60680-03
CAAAJ60680-MD
Hydroxytacrine maleate salt
Hydroxytacrine maleate salt
Cholinesterase inhibitor. Potential Alzheimer's disease therapeutic
Solid
Applications: Cholinesterase inhibitor. Potential Alzheimer's disease therapeutic
Formula:
C₁₇H₁₈N₂O₅ Melting Pt: 171…173 °C Storage Temperature: Freezer |
MDL Number:
MFCD00069287 CAS Number: 118909-22-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...