4-Thiouridine ≥98%
Supplier: Thermo Scientific Chemicals
Synonyms:
1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-4-thioxo-3,4-dihydropyrimidin-2(1H)-one
J60679-MC
J60679-MD
CAAAJ60679-MCEA
1053.45
CAD
CAAAJ60679-MC
AAJ60679-MD
4-Thiouridine ≥98%
4-Thiouridine
Crystalline
Purity: 98+%
CAS No.: 13957-31-8
Molecular Formula: C9H12N2O5S
Formula Weight: 260.27
Storage Temperature: -30°C to -10°C
Physical Form: Crystalline
MDL No.: MFCD00006538
Formula:
C₉H₁₂N₂O₅S MW: 260.264 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00006538 CAS Number: 13957-31-8 EINECS: 237-735-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...