Adenosine-5'-diphosphate monopotassium salt dihydrate 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
5'-ADP-K dihydrate, Potassium (2R,3R,4S,5R)-2-(6-amino-9H-purin-9-yl)-4-hydroxy-5-(((hydroxy(phosphonooxy)phosphoryl)oxy)methyl)tetrahydrofuran-3-olate dihydrate, ADP-K dihydrate
J60672-03
J60672-06
CAAAJ60672-03EA
186.93
CAD
CAAAJ60672-03
CAAAJ60672-06
Adenosine-5'-diphosphate monopotassium salt dihydrate 99%
Adenosine-5'-diphosphate monopotassium salt dihydrate
Crystalline powder
Purity: 99%
CAS No.: 72696-48-1
Molecular Formula: C10H14KN5O10P2.2H2O
Formula Weight: 501.33 (465.29anhy)
Storage Temperature: -30°C to -10°C
Physical Form: Crystalline powder
Appearance: White
MDL No.: MFCD00066472
Formula:
C₁₀H₁₄KN₅O₁₀P₂·2H₂O MW: 501.33 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00150944 CAS Number: 72696-48-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...