Kanamycin sulfate cell culture reagent
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Kanamycin Monosulfate, (2R,3S,4S,5R,6R)-2-(Aminomethyl)-6-(((1R,2R,3S,4R,6S)-4,6-diamino-3-(((2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)tetrahydro-2H-pyran-3,4,5-triol sulfate, Kanamycin A, Kanamycin Monosulphate, (2R,3S,4S,5R,6R)-2-(Aminomethyl)-6-(((1R,2R,3S,4R,6S)-4,6-diamino-3-(((2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)tetrahydro-2H-pyran-3,4,5-triol sulphate
Powder
CAS No.: 25389-94-0
Molecular Formula: C18H36N4O11.H2SO4
Formula Weight: 582.58
Physical Form: Powder
Appearance: White to off-white
Solubility: Soluble in water at 10mg/ml
MDL No.: MFCD00070253
Formula:
C₁₈H₃₈N₄O₁₅S MW: 582.6 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00070253 CAS Number: 25389-94-0 EINECS: 246-933-9 Merck Index: 14,5281 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...