Dihydroergocristine methanesulfonate

Supplier: Thermo Scientific Chemicals

Synonyms: Dihydroergocristine mesylate

J60638-03 J60638-EX3
CAAAJ60638-03EA 1340.79 CAD
CAAAJ60638-03 CAAAJ60638-EX3
Dihydroergocristine methanesulfonate
Dihydroergocristine methanesulfonate

5-HT receptor antagonist; partial agonist at adrenergic and dopaminergic receptors


Powder


5-HT receptor antagonist; partial agonist at adrenergic and dopaminergic receptors

Formula: C₃₆H₄₅N₅O₈S
Storage Temperature: Ambient
MDL Number: MFCD00153792
CAS Number: 24730-10-7
EINECS: 246-434-6
Merck Index: 14,3653

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR