DNAzol® Reagent, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J60637-AD
J60637-AE
J60637-AH
CAAAJ60637-ADEA
394.42
CAD
CAAAJ60637-AD
CAAAJ60637-AE
CAAAJ60637-AH
DNAzol® Reagent, Thermo Scientific
Nucleic Acid Reagents
Nucleic Acid Purification Kits and Reagents
DNAzol® Reagent is used as an electrophoretic reagent for the isolation of genomic DNA from solid and liquid samples of animal and plant origin.
- Liquid, soluble in water
- Store at ambient temperature