Zolmitriptan

Supplier: Thermo Scientific Chemicals

J60616-03 J60616-MC
CAAAJ60616-03EA 1242.09 CAD
CAAAJ60616-03 CAAAJ60616-MC
Zolmitriptan
Zolmitriptan

A serotonin 5HT1D-receptor agonist


Crystalline powder


Applications: A serotonin 5HT1D-receptor agonist

Formula: C₁₆H₂₁N₃O₂
Melting Pt: 136…141 °C
Storage Temperature: Ambient
MDL Number: MFCD00871503
CAS Number: 139264-17-8
Merck Index: 14,10189

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR