Iota carrageenan

Supplier: Thermo Scientific Chemicals

J60603-22 J60603-30
CAAAJ60603-22EA 122.45 CAD
CAAAJ60603-22 CAAAJ60603-30
Iota carrageenan
Iota carrageenan

Forms soft gels in the presence of calcium ions


Powder


Applications: Forms soft gels in the presence of calcium ions

Storage Temperature: Ambient
MDL Number: MFCD00151512
CAS Number: 9062-07-1
EINECS: 232-949-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR