Fasudil dihydrochloride ≥99%

Supplier: Thermo Scientific Chemicals

J60594-03 J60594-MD J60594-ME
CAAAJ60594-03EA 3059.35 CAD
CAAAJ60594-03 CAAAJ60594-MD CAAAJ60594-ME
Fasudil dihydrochloride ≥99%
Fasudil dihydrochloride

A vasodilator agent which inhibits vascular smooth muscle contraction by acting as an intracellular calcium antagonist


Powder


Applications: A vasodilator agent which inhibits vascular smooth muscle contraction by acting as an intracellular calcium antagonist

Formula: C₁₄H₁₇N₃O₂S·2HCl
Storage Temperature: Freezer
MDL Number: MFCD00153805
CAS Number: 203911-27-7
Merck Index: 14,3942

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR