Naltrexone hydrochloride

Supplier: Thermo Scientific Chemicals
Warning

J60590-03 J60590-MD
CAAAJ60590-03EA 1468.99 CAD
CAAAJ60590-03 CAAAJ60590-MD
Naltrexone hydrochloride
Naltrexone hydrochloride

A congener of naloxone and also an opioid antagonist


Powder


Applications: A congener of naloxone and also an opioid antagonist

Formula: C₁₉H₂₁NO₄·HCl
Storage Temperature: Refrigerator
MDL Number: MFCD00069324
CAS Number: 16676-29-2
EINECS: 240-723-0
Merck Index: 14,6363

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR