Potassium phosphate, 0.5 M buffer soln., pH 7.0, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J60584-AK J60584-AP
CAAAJ60584-AKEA 175.14 CAD
CAAAJ60584-AK CAAAJ60584-AP
Potassium phosphate, 0.5 M buffer soln., pH 7.0, Thermo Scientific
Buffers Biological Buffers

Liquid


CAS No.: 7778-77-0
Physical Form: Liquid
MDL No.: MFCD00011401

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR