Doxycycline Hyclate

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Doxycycline Hydrochloride Hemiethanolate Hemihydrate

J60579-14 J60579-22
CAAAJ60579-14EA 187.18 CAD
CAAAJ60579-14 CAAAJ60579-22
Doxycycline Hyclate
Doxycycline hyclate

Inhibits matrix metalloproteinases at subantimicrobial doses


Powder


Applications: Inhibits matrix metalloproteinases at subantimicrobial doses

Formula: C₂₂H₂₄N₂O₈·HCl·0.5H₂O·0.5C₂H₆O
Storage Temperature: Refrigerator
MDL Number: MFCD07357237
CAS Number: 24390-14-5
Merck Index: 14,3440

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR