CAPS, 0.5 M Buffer Solutions, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J60569-AK
J60569-AP
J62446-AK
J62446-AP
J63138-AK
J63138-AP
J63749-AK
CAAAJ60569-AKEA
175.14
CAD
CAAAJ60569-AK
CAAAJ60569-AP
CAAAJ60776-AE
CAAAJ60776-AK
CAAAJ62446-AK
CAAAJ62446-AP
CAAAJ63138-AK
CAAAJ63138-AP
CAAAJ63749-AK
CAAAJ63749-AP
CAPS, 0.5 M Buffer Solutions, Thermo Scientific
Buffers
Biological Buffers
Liquid
CAS No.: 1135-40-6
MDL No.: MFCD00003837
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...