Sodium phosphate, 0.2 M buffer soln., pH 9.0, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J60565-AP J60565-K2
CAAAJ60565-APEA 135.99 CAD
CAAAJ60565-AP CAAAJ60565-K2
Sodium phosphate, 0.2 M buffer soln., pH 9.0, Thermo Scientific
Buffers Biological Buffers

Liquid


Physical Form: Liquid
MDL No.: MFCD00146206

Order Now

SPECIFICATIONS

Environmentally Preferable
Physical form
MDL No. MFCD00146206

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR