Orange G 6X loading dye, glycerol based

Supplier: Thermo Scientific Chemicals

Synonyms: Disodium 7-hydroxy-8-phenylazonaphthalene-1,3-disulphonate, Wool Orange 2G, Orange G sodium salt, CI 16230, Acid Orange 10, Disodium 7-hydroxy-8-phenylazonaphthalene-1,3-disulfonate

J60562-AC J60562-AD
CAAAJ60562-ACEA 121.41 CAD
CAAAJ60562-AC CAAAJ60562-AD
Orange G 6X loading dye, glycerol based
Orange G

DNA loading dye used to prepare markers and samples for loading on agarose or polyacrylamide gels in molecular biology protocols.


  • Recommended for small molecules


The 6X Orange DNA Loading Dye is used to prepare DNA markers and samples for loading on agarose or polyacrylamide gels. It is recommended for the analysis of small DNA molecules.


Contains: 0.25% Orange-G and 30% glycerol in autoclaved DI water

Formula: C₁₆H₁₀N₂Na₂O₇S₂
MW: 452.38 g/mol
Melting Pt: 141
Storage Temperature: Ambient
MDL Number: MFCD00012457
CAS Number: 1936-15-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR