SSC (20X), RNAse Free, Thermo Scientific
Supplier: Thermo Scientific
J60561-AP J60561-AP J60561-AK J60561-AK
CAAAJ60561-APEA 323.81 CAD
AAJ60561-AK CAAAJ60561-AP AAJ60561-AP CAAAJ60561-AK
SSC (20X), RNAse Free, Thermo Scientific
Buffers Electrophoresis Buffers Hybridisation Buffers
SSC solutions are commonly used in nucleic acid hybridization techniques at concentrations ranging from 0.1X to 20X depending on the application.
  • Contains 0.3 M sodium citrate, 3.0 M sodium chloride
These are used to facilitate transfer of nucleic acids to membranes in Southern and Northern blotting, dot blotting, and colony and plaque lifts.