Adenosine-2',3'-cyclic monophosphate sodium salt 97%
Supplier: Thermo Scientific Chemicals
J60558-MC
J60558-MD
CAAAJ60558-MCEA
507.43
CAD
CAAAJ60558-MC
CAAAJ60558-MD
CAAAJ60558-ME
Adenosine-2',3'-cyclic monophosphate sodium salt 97%
Adenosine-2',3'-cyclic monophosphate sodium salt
Powder
Purity: 97%
CAS No.: 37063-35-7
Molecular Formula: C10H11N5NaO6P
Formula Weight: 351.19
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
Melting Point: 241-243°
MDL No.: MFCD00005757
Formula:
C₁₀H₁₁N₅NaO₆P MW: 351.19 g/mol Melting Pt: 241…243 °C Storage Temperature: Freezer |
MDL Number:
MFCD00005757 CAS Number: 37063-35-7 EINECS: 253-328-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...