3-Indoxyl phosphate disodium salt
Supplier: Thermo Scientific Chemicals
Synonyms:
3-Indolyl phosphate disodium salt
J60552-03
J60552-MD
CAAAJ60552-03EA
641.62
CAD
CAAAJ60552-03
CAAAJ60552-MD
3-Indoxyl phosphate disodium salt
3-Indoxyl phosphate disodium salt
A histochemical substrate for alkaline phosphatase
Powder
Applications: A histochemical substrate for alkaline phosphatase
Formula:
C₈H₆NNa₂O₄P MW: 257.09 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00040646 CAS Number: 3318-43-2 EINECS: 222-014-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...