Vincamine 98%
Supplier: Thermo Scientific Chemicals
J60545-03
J60545-06
CAAAJ60545-03EA
315.26
CAD
CAAAJ60545-03
CAAAJ60545-06
Vincamine 98%
Vincamine
A vasodilator
Powder
Applications: A vasodilator
Formula:
C₂₁H₂₆N₂O₃ Melting Pt: 232 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00078054 CAS Number: 1617-90-9 EINECS: 216-576-3 Merck Index: 14,9983 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...