EPPS, 0.2 M Buffer Solutions, Thermo Scientific

Supplier: Thermo Scientific
J61538-AE J61538-AK J60511-AE J61538-AK J61296-AE J61296-AK J61476-AE J61476-AE J61538-AE J61296-AE J60511-AK J61476-AK J61476-AK J60511-AE J61296-AK J60511-AK
CAAAJ60511-AKEA 267.13 CAD
CAAAJ60511-AK AAJ60511-AK AAJ60511-AE AAJ61476-AK AAJ61538-AK CAAAJ60511-AE CAAAJ61538-AK CAAAJ61296-AK CAAAJ61296-AE AAJ61476-AE CAAAJ61538-AE AAJ61296-AK CAAAJ61476-AE AAJ61538-AE AAJ61296-AE CAAAJ61476-AK
EPPS, 0.2 M Buffer Solutions, Thermo Scientific
Buffers Biological Buffers
Liquid

CAS No.: 16052-06-5
Physical Form: Liquid
MDL No.: MFCD00006160
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR