TRIS-buffered saline (TBS, 20X), with 2% Tween 20, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J60497-K2 J60497-K2 J60497-K3
AAJ60497-K2EA 249.64 CAD
AAJ60497-K2 CAAAJ60497-K2 CAAAJ60497-K3
TRIS-buffered saline (TBS, 20X), with 2% Tween 20, Thermo Scientific
Buffers Electrophoresis Buffers Washing Buffers

Liquid


Physical Form: Liquid
MDL No.: MFCD00678198

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR