BICINE, 0.5 M Buffer Solutions, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J60494-AE J60494-AK J61632-AE J61632-AK J63924-AE J63924-AK
CAAAJ60494-AEEA 135.96 CAD
CAAAJ60494-AE CAAAJ60494-AK CAAAJ61632-AE CAAAJ61632-AK CAAAJ62838-AE CAAAJ62838-AK CAAAJ63924-AE CAAAJ63924-AK
BICINE, 0.5 M Buffer Solutions, Thermo Scientific
Buffers Biological Buffers

N,N-(2-Dihydroxyethyl)glycine (BICINE) buffer solutions.


CAS No.: 150-25-4
MDL No.: MFCD00004295


BICINE is used for the preparation of stable substrate solution for serum guanase determination. The use of bicine in a thin layer ion exchange chromatography method for protein resolution has been published. Bicine has been utilized in peptide and protein crystallization. A kinetic study of a quaternary transition-state analogue complex of creatine kinase used bicine in the reaction buffer. A multiphasic buffer system for SDS-PAGE of proteins and peptides that incorporates bicine has been described.


Caution: Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR